Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ RTX™-5 Capillary Columns, 60m or Longer
SDP

Catalog No. 06708648 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
60 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Particle Size:
0.1 μm
0.25 μm
0.5 μm
1 μm
1.5 μm
3 μm
5 μm
Includes:
5m Integra-Guard Column
Cage
Integra guard column
n.a.
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Inner Particle Size Includes
06-708-648 60 m 0.32 mm 0.5 μm -
06-708-601 60 m 0.32 mm 0.25 μm 5m Integra-Guard Column
06-708-536 60 m 0.25 mm 0.1 μm Integra guard column
06-708-594 60 m 0.25 mm 0.25 μm -
06-708-602 60 m 0.32 mm 0.25 μm Integra guard column
06-708-646 60 m 0.32 mm 0.5 μm Integra guard column
06-708-647 60 m 0.32 mm 0.5 μm Integra guard column
06-708-695 60 m 0.25 mm 1 μm Integra guard column
06-708-700 60 m 0.32 mm 1 μm Integra guard column
06-708-704 60 m 0.53 mm 1 μm n.a.
06-708-727 60 m 0.53 mm 1.5 μm Cage
06-708-747 60 m 0.53 mm 5 μm Integra guard column
06-708-760 60 m 0.32 mm 3 μm Integra guard column
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 06-708-648 Supplier Restek™ Supplier No. 10242138

General-purpose, fused silica columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, semivolatiles.

The diphenyl dimethyl polysiloxane stationary phase is the most popular GC stationary phase and is used in a wide variety of applications. All residual catalysts and low molecular weight fragments are removed from the Rtx-5 polymer, providing a tight monomodal distribution and extremely low bleed.

  • Low-polarity phase
  • Crossbond diphenyl dimethyl polysiloxane
  • Equivalent to USP G27 and G36 phases

Note
DB-5MS is a silarylene-based polymer, similar to Rxi-5Sil MS.

Order Info

Custom lengths and film thicknesses available.

Specifications

For Use With (Application) Drug Analysis, Solvent Impurities Analysis, Pesticide Analysis, Hydrocarbon Analysis, PCB Congener Analysis, Essential Oils, Semivolatiles Analysis
Particle Size 0.5 μm
Film Thickness 0.5 μm
Length (Metric) 60 m
Material Stainless Steel
Packing Material Rtx™-5
Polarity Low polar
Coating Fused Silica
USP Type G27, G36
Stationary Phase 5% Diphenyl/95% Dimethyl Polysiloxane
Diameter (Metric) Inner 0.32 mm
Max. Temperature (Metric) 350°C
Temperature Range (Metric) -60°C to +330/350°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.