Learn More
Description
- Especially well-suited for analysis of active, high molecular weight compounds with sensitive GC/MS systems including ion trap detectors
- Temperature range 30° to 360°C for 15m and 30m lengths: longer columns may have a slightly reduced maximum temperature
- Well-suited for U.S. EPA Method 525 for analysis of semivolatile compounds in drinking water
Pesticides, PCB cogeners
Specifications
Specifications
| For Use With (Application) | Pesticide Analysis, PCB Congener Analysis, PAHs Analysis |
| Particle Size | 1 μm |
| Film Thickness | 1 μm |
| Length (Metric) | 30 m |
| Polarity | Low polar |
| Coating | Fused Silica |
| Stationary Phase | Rxi™-XLB |
| Diameter (Metric) Inner | 0.25 mm |
| Temperature Range (Metric) | 30°C to 340/360°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
