Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ SPI Liners for Varian, With Buffer
SDP

Catalog No. 06718102 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25/Pk.
5/Pk.
Each
Deactivation:
Intermediate Polarity (IP)
Wool
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Deactivation
06-718-102 5/Pk. Wool
06-711-147 Each Intermediate Polarity (IP)
06-718-095 5/Pk. Intermediate Polarity (IP)
06-718-104 25/Pk. Intermediate Polarity (IP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-718-102 Supplier Restek™ Supplier No. 20851213.5

For Bruker/Varian GCs equipped with SPI inlets.

  • Geometry: SPI with Buffer
  • Material: Borosilicate Glass
  • Deactivation: Intermediate Polarity (IP)
  • Liner ID: 2.4mm
  • Length: 54mm
  • OD: 4.6mm

Specifications

Product Type SPI Liner
Packing Material Wool
Length (Metric) 54 mm
Quantity 5/Pk.
Includes Buffer
Deactivation Wool
Geometry SPI with Buffer
Diameter (Metric) Inner 2.4 mm
Diameter (Metric) Outer 4.6 mm
For Use With (Equipment) Bruker/Varian GCs Equipped with SPI Inlets
Liner Material Borosilicate Glass
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.