Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ SPI Liners for Varian, Without Buffer
SDP

Catalog No. 06717921 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Diameter (Metric) Inner:
0.53 mm
0.80 mm
Quantity:
25/Pk.
5/Pk.
Each
Deactivation:
Intermediate Polarity (IP)
Siltek
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner Quantity Deactivation
06-717-921 0.53 mm 5/Pk. Siltek
06-711-011 0.53 mm Each Intermediate Polarity (IP)
06-711-015 0.80 mm Each Intermediate Polarity (IP)
06-717-922 0.53 mm 25/Pk. Intermediate Polarity (IP)
06-717-923 0.53 mm 25/Pk. Siltek
06-717-930 0.80 mm 25/Pk. Intermediate Polarity (IP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 06-717-921 Supplier Restek™ Supplier No. 20776214.5

For Bruker/Varian GCs equipped with SPI inlets.

  • Geometry: Straight
  • Material: Borosilicate Glass
  • OD: 4.6mm
  • Length: 54mm

Specifications

Product Type SPI Straight Inlet Liner
Length (Metric) 54 mm
Quantity 5/Pk.
Deactivation Siltek
Geometry Straight
Diameter (Metric) Inner 0.53 mm
Diameter (Metric) Outer 4.6 mm
For Use With (Equipment) Bruker/Varian GCs Equipped with SPI Inlets
Liner Material Borosilicate Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.