Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ SPI Liners for Varian, Without Buffer

For Bruker/Varian GCs equipped with SPI inlets.
Supplier: Restek™ 20779
Description
- Geometry: Straight
- Material: Borosilicate Glass
- OD: 4.6mm
- Length: 54mm
Specifications
54 mm | |
5/Pk. | |
Straight | |
4.6 mm | |
Borosilicate Glass |
SPI Straight Inlet Liner | |
Intermediate Polarity (IP) | |
0.80 mm | |
Bruker/Varian GCs Equipped with SPI Inlets |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction