Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ SPI Liners for Varian, Without Buffer

For Bruker/Varian GCs equipped with SPI inlets.
$209.00 - $272.00
Specifications
Length (Metric) | 54 mm |
---|---|
Product Type | SPI Straight Inlet Liner |
Geometry | Straight |
Diameter (Metric) Outer | 4.6 mm |
For Use With (Equipment) | Bruker/Varian GCs Equipped with SPI Inlets |
Catalog Number | Mfr. No. | Deactivation | Diameter (Metric) Inner | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Deactivation | Diameter (Metric) Inner | Quantity | Price | Quantity & Availability | |||
06-717-924
![]() |
Restek™
20779 |
Intermediate Polarity (IP) | 0.80 mm | 5/Pk. |
Pack of 5 for $209.00
|
|
|||
06-717-929
![]() |
Restek™
20779214 |
Siltek | 0.80 mm | 5/Pk. |
Pack of 5 for $272.00
|
|
|||
06-711-011
![]() |
Restek™
20775 |
Intermediate Polarity (IP) | 0.53 mm | Each | N/A | N/A | N/A | ||
06-711-015
![]() |
Restek™
20778 |
Intermediate Polarity (IP) | 0.80 mm | Each | N/A | N/A | N/A | ||
06-717-921
![]() |
Restek™
20776214.5 |
Siltek | 0.53 mm | 5/Pk. | N/A | N/A | N/A | ||
06-717-922
![]() |
Restek™
20777 |
Intermediate Polarity (IP) | 0.53 mm | 25/Pk. | N/A | N/A | N/A | ||
06-717-923
![]() |
Restek™
20777214.25 |
Siltek | 0.53 mm | 25/Pk. | N/A | N/A | N/A | ||
06-717-930
![]() |
Restek™
20780 |
Intermediate Polarity (IP) | 0.80 mm | 25/Pk. | N/A | N/A | N/A | ||
Description
- Geometry: Straight
- Material: Borosilicate Glass
- OD: 4.6mm
- Length: 54mm
Specifications
54 mm | |
Straight | |
Bruker/Varian GCs Equipped with SPI Inlets |
SPI Straight Inlet Liner | |
4.6 mm | |
Borosilicate Glass |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title