Learn More
Description
- Material: Borosilicate Glass
- OD: 6.3mm
- Length: 72mm
Specifications
Specifications
| Product Type | Split Inlet Liner |
| Length (Metric) | 72 mm |
| For Use With (Application) | For High and Low MW Compounds |
| Geometry | Cup with Top Taper |
| Deactivation | Intermediate-Polarity |
| Quantity | Each |
| Diameter (Metric) Inner | 4 mm |
| Diameter (Metric) Outer | 6.3 mm |
| For Use With (Equipment) | For Bruker/Varian GCs |
| Liner Material | Borosilicate Glass |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
