Learn More
Description
- Acidic functionality incorporated into the polymer structure
- Capable of analyzing acidic compounds without derivatization, significantly reducing adsorption of acids to increase sample capacity for volatile-free acids
- Ideal for analyzing distilled liquor and other alcoholic beverages
- Improved peak shapes for high molecular weight acids
- Compatible with some inorganic acids depending on the volatility
- Resistant to oxidative damage
- Equivalent to USP G25, G35 phases
- Do not rinse with water
Specifications
Specifications
| For Use With (Application) | Free (Underivatized) Acids, and Various Inorganic Acids |
| Film Thickness | 0.5 μm |
| Length (Metric) | 15 m |
| Polarity | Polar |
| Coating | Fused Silica |
| Description | Resistant to oxidative damage |
| USP Type | G25, G35 |
| Stationary Phase | Stabilwax™-DA |
| Diameter (Metric) Inner | 0.53 mm |
| Max. Temperature (Metric) | 250°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
