Learn More
Restek™ Tubing Dressing Tool
SDP
Catalog No. 06710542 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Replacement Needle
Tubing Dressing Tool
Diameter (English) Inner:
1/16 in.
1/8 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Diameter (English) Inner
06-710-542 Tubing Dressing Tool 1/16 in.
06-710-543 Replacement Needle 1/16 in.
06-710-545 Replacement Needle 1/8 in.
06-710-544 Tubing Dressing Tool 1/8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-710-542 Supplier Restek™ Supplier No. 20188
Convenient tool opens stainless steel tubing bore and removes burrs.
Tube Dressing Tool is also Included with the SSI TC-20 Tube Cutting Machine.
Specifications
Type Tubing Dressing Tool
Diameter (English) Inner 1/16 in.
Packaging Quantity 1 Each