Learn More
Restek™ UltraShield™ UHPLC PreColumn Filter
SDP
Catalog No. 06715981 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
1 Each
10/Pk.
5/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
06-715-981 1 Each
06-715-983 10/Pk.
06-715-982 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-715-981 Supplier Restek™ Supplier No. 24995
Add to Cart
Add to Cart
Cost-effective protection for UHPLC systems
  • In-line design installs easily with standard fittings
  • Reliable way to extend column lifetime
  • Leak-tight to 15,000 psi (1034 bar)
  • 0.5μm titaniun filter in stainless steel body withh PEEK ferrule
Specifications
For Use With (Application) For Protection From Harmful Effects of Sample and Mobile Phase Particulates on UHPLC Columns
Material Stainless Steel, PEEK (Ferrule)
Packaging Quantity 1 Each
Type UHPLC PreColumn Filter