Learn More
Uniliner Liners for 1/4-Inch Packed Injection Port Conversion from Restek™
For 0.25 in. packed injection port conversion.
Supplier: Restek Corporation 20305
Description
- Reduces solvent tailing
- 0.53mm ID version can be used in the direct (DI) or on-column (OC) injection mode
- Incorporates a gentle taper that seals the column and reduces dead volume in direct injection mode
Specifications
Uniliner liner, Direct Injection/On-Column | |
5/Pk. | |
0.5 cm |
Uniliner; Direct Injection/On-Column; 85mm long | |
0.53 mm | |
0.53 mm I.D. On-Column Injection Mode, 0.25 in. Packed Injection Port Conversion, Large Buffer Volume Chamber 85 mm L, for ≤4 μL Injections |