Learn More
Description
- 300 series stainless steel
- Includes stainless steel ferrules
- Zero-Dead-Volume Valco Internal Tee connects three lines
- Zero-Dead-Volume Valco Internal Cross connects four lines
- Union Bore: 0.25mm
Specifications
Specifications
| Type | Tee |
| Diameter (Metric) Inner | 0.25 mm |
| Thread Size | 0.25mm bore |
| Description | Internal Tee |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
