Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Restriction Enzyme BglII
Click to view available options
Quantity:
10,000 U
Description
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
Specifications
Specifications
Concentration | 10 U/μL |
Content And Storage | -30°C to -10°C |
Source | Bacillus globigii |
Quantity | 10,000 U |
Product Type | Restriction Enzyme BglII |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction