Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Restriction Enzymes - EcoR I Blue/White Cloning Qualified (H)

Catalog No. PRR4014 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
15,000 U
25,000 U
Concentration:
12 U/μL
40 to 80 U/μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
PR-R4014 25,000 U 40 to 80 U/μL
PR-R6017 15,000 U 12 U/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PR-R4014 Supplier Promega Supplier No. R4014
Add to Cart
Add to Cart
This item is not returnable. View return policy

Capable of digesting DNA in 15 minutes or less. (G/AATT C C TTAA\G)

  • Capable of digesting DNA in 15 minutes or less
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Available at high concentration, containing 25,000 and 50,000 units of EcoRI at a concentration of 40-80u/μL

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Specifications

Concentration 40 to 80 U/μL
Components 1 x 1mL 10X Buffer H, 1 x 1250μL Bovine Serum Albumin Acetylated, 1 x 25000U EcoRI (HC)
Incubator Temperature 37°C
pH 7.4
For Use With (Application) Capable of digesting DNA in <15 min.
Storage Buffer 10mM Tris HCl (pH 7.4), 400mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 0.15% Triton™ X-100, 50% Glycerol
Content And Storage -30°C to -10°C
Quantity 25,000 U
Cut Site G.AATTC
Product Type EcoRI (HC)
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.