Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Restriction Enzymes - EcoR I Blue/White Cloning Qualified (H)

Catalog No. p-2387073
Click to view available options
Quantity:
15,000 U
25,000 U
5000 U
Concentration:
12 U/μL
40 to 80 U/μL

Capable of digesting DNA in 15 minutes or less. (G/AATT C C TTAA\G)

  • Capable of digesting DNA in 15 minutes or less
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Available at high concentration, containing 25,000 and 50,000 units of EcoRI at a concentration of 40-80u/μL

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Specifications

Concentration 12 U/μL
Components 1 x 1mL 10X Buffer H, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 15000U EcoRI
Incubator Temperature 37°C
pH 7.4
For Use With (Application) Capable of digesting DNA in <15 min.
Storage Buffer 10mM Tris HCl (pH 7.4), 400mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 0.15% Triton™ X-100, 50% Glycerol
Content And Storage -30°C to -10°C
Quantity 15,000 U
Cut Site G.AATTC
Product Type EcoRI
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.