Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Restriction Enzymes - Hind III Blue/White Cloning Qualified (E)

Catalog No. PRR6041 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
15,000 U
5000 U
Concentration:
10 U/μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
PR-R6041 5000 U 10 U/μL
PR-R6045 15,000 U 10 U/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PR-R6041 Supplier Promega Supplier No. R6041
Add to Cart
Add to Cart
This item is not returnable. View return policy

Capable of digesting DNA in 15 minutes or less. Star activity may be observed in the presence of Mn2+. (A/AGCT T T TCGA\A)

  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Specifications

Concentration 10 U/μL
Components 1 x 1mL 10X Buffer E, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 5000U HindIII, 1 x 0.25mL Multi-Core™ 10X Buffer
Incubator Temperature 37°C
For Use With (Application) Provides a higher level of quality control for Enzymes used in cloning applications
Storage Buffer 10mM Tris HCl (pH 7.4), 250mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Content And Storage -30°C to -10°C
Quantity 5000 U
Cut Site A.AGCTT TTCGA.A
Product Type HindIII
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.