Learn More
Promega Restriction Enzymes - Not I Blue/White Cloning and Genome Qualified (D)
Shop All Promega Corporation Products
Description
- Capable of digesting DNA in 15 minutes or less
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
- Genome qualified to ensure optimal performance in genomic analysis applications
- High concentration product no longer available
‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
Specifications
| Concentration | 10 U/μL |
| Components | 1 x 1mL 10X Buffer D, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 200U NotI, 1 x 0.25mL Multi-Core™ 10X Buffer |
| Incubator Temperature | 37°C |
| pH | 7.4 |
| For Use With (Application) | Capable of digesting DNA in <15 min. |
| Storage Buffer | 10mM Tris HCl (pH 7.4), 500mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 0.1% Triton™ X-100, 50% Glycerol |
| Content And Storage | -30°C to -10°C |
| Quantity | 200 U |
| Cut Site | GC.GGCCGC CGCCGG.CG |
| Product Type | NotI |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.