Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ret Antibody (RET/2795), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB012075 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB012075 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB012075 Supplier Novus Biologicals Supplier No. NBP279897AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ret Monoclonal specifically detects Ret in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Ret
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RET/2795
Conjugate Alexa Fluor 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence
Gene Symbols RET
Host Species Mouse
Immunogen Recombinant fragment (around aa 702-848) of human Ret protein (exact sequence is proprietary) (Uniprot: P07949)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Oncogenes, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.