Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Retinol Binding Protein RBP Antibody (rRBP1/872), HRP, Novus Biologicals™
SDP

Catalog No. NB383481 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383481 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383481 Supplier Novus Biologicals Supplier No. NBP308941H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Retinol Binding Protein RBP Monoclonal antibody specifically detects Retinol Binding Protein RBP in Human, Mouse, Rat, Goat, Chimpanzee, Monkey, Rabbit samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Retinol Binding Protein RBP
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rRBP1/872
Conjugate HRP
Formulation PBS
Gene Alias C, Cellular retinol-binding protein, CRABP-I, CRBP1CRBP-I, CRBPCellular retinol-binding protein I, CRBPI, RBPC, retinol binding protein 1, cellular, retinol-binding protein 1, retinol-binding protein 1, cellular
Host Species Mouse
Immunogen Recombinant full-length human Retinol Binding Protein RBP protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5947
Target Species Human, Mouse, Rat, Goat, Chimpanzee
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.