Learn More
Retinol Binding Protein RBP Antibody (SPM442), HRP, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | RBP4/Retinol-Binding Protein 4 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | SPM442 |
| Conjugate | HRP |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Gene Alias | interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma |
| Gene Symbols | RBP4 |
| Host Species | Mouse |
| Immunogen | Retinol binding protein purified from human plasma |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.