Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Retractable Badge Holder

Catalog No. 19103141 Shop All Brady Corporation Products
Click to view available options
Color:
Black
Blue
Gray
Red
White
This item is not returnable. View return policy
This item is not returnable. View return policy

Stop losing ID cards. The Brady™ Retractable Badge Holders keep your ID, access or security card near, while allowing the range to scan or swipe as needed.

  • ID tag
  • Lifetime guarantee
  • Ready-to-roll, full factory assembly

  • Ideal alternative to bull dog clips which may damage clothing
  • Clip attaches to belt or pocket while the heavy-duty nylon cable with fastener attaches to badge

Specifications

Color Gray
For Use With (Equipment) ID badges, access and security cards
Includes 1/Each
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.