Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Retractable Badge Holder
Shop All Brady Corporation Products
Click to view available options
Color:
Black
Blue
Gray
Red
White
Description
- ID tag
- Lifetime guarantee
- Ready-to-roll, full factory assembly
- Ideal alternative to bull dog clips which may damage clothing
- Clip attaches to belt or pocket while the heavy-duty nylon cable with fastener attaches to badge
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | ID badges, access and security cards |
| Includes | 1/Each |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction