Learn More
RETSCH ZM 200 Ultra Centrifugal Mill, Push-Fit Rotors
Catalog No. 084164 Shop All RETSCH Inc. Products
Click to view available options
Type:
12 Tooth SS - Medium Coarse Materials
6 Tooth Rotor
For fine grinding of soft to medium-hard and fibrous materials.
  • Accessory for use with ZM 200 Ultra Centrifugal Mill
  • Wear resistant coating
Specifications
Material Stainless Steel
Type 12 Tooth SS - Medium Coarse Materials
Max. Speed 6,000 to 18,000 rpm
Max. Tube Diameter 9.9 cm
For Use With (Equipment) Centrifugal Mill
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.