Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rev-erb A alpha/NR1D1 Rabbit anti-Human, Mouse, Rat, Clone: 9I2H1, Novus Biologicals™
SDP

Catalog No. NB166891 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166891 100 μg
NB166892 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166891 Supplier Novus Biologicals Supplier No. NBP315898100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rev-erb A alpha/NR1D1 Monoclonal antibody specifically detects Rev-erb A alpha/NR1D1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Rev-erb A alpha/NR1D1
Applications Western Blot
Classification Monoclonal
Clone 9I2H1
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EAR-1, EAR1nuclear receptor subfamily 1 group D member 1, HREV, nuclear receptor subfamily 1, group D, member 1, Rev-erbA-alpha, Rev-ErbAalpha, THRA1, THRALrev-erbA-alpha, V-erbA-related protein 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 280-360 of human Rev-erb A alpha/NR1D1 (NP_068370.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9572
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.