Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rev-erb beta/NR1D2 Antibody (OTI5E10), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP27384A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27384A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27384A70 Supplier Novus Biologicals Supplier No. NBP273854AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rev-erb beta/NR1D2 Monoclonal specifically detects Rev-erb beta/NR1D2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Rev-erb beta/NR1D2
Applications Western Blot
Classification Monoclonal
Clone OTI5E10
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias BD73, EAR-1r, HZF2, nuclear receptor subfamily 1 group D member 2, nuclear receptor subfamily 1, group D, member 2, Orphan nuclear hormone receptor BD73, rev-erba-alpha-related receptor, rev-erb-beta, RVR, V-erbA-related protein 1-related
Gene Symbols NR1D2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-294 of human Rev-erb beta/NR1D2 (NP_005117) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9975
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.