Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rex-1/ZFP42 Antibody (OTI3H9), HRP, Novus Biologicals™
SDP

Catalog No. NBP273855H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273855H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273855H Supplier Novus Biologicals Supplier No. NBP273855H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rex-1/ZFP42 Monoclonal specifically detects Rex-1/ZFP42 in Human samples. It is validated for Western Blot.

Specifications

Antigen Rex-1/ZFP42
Applications Western Blot
Classification Monoclonal
Clone OTI3H9
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias FLJ20086, KIAA1584, MGC21637, MGC61687, SUHW4, Suppressor of hairy wing homolog 4, zinc finger protein 280D, Zinc finger protein 634, ZNF634suppressor of hairy wing homolog 4 (Drosophila)
Gene Symbols ZFP42
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-235 of human Rex-1/ZFP42 (NP_777560) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 132625.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.