Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501728790 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8790 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8790 Supplier Proteintech Group Inc Supplier No. 135031AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Seems to have no detectable effect on transcription elongation in vitro. Tamura K., Biochem. Biophys. Res. Commun. 309:189-195(2003). Hirosawa M., DNA Res. 6:329-336(1999). The MGC Project Team; Genome Res. 14:2121-2127(2004).

Specifications

Antigen REX1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene REXO1
Gene Accession No. Q7TT28, Q8N1G1
Gene Alias EloA BP1, ELOABP1, Elongin A binding protein 1, KIAA1138, REX1, REXO1, RNA exonuclease 1 homolog, TCEB3BP1
Gene Symbols Rexo1
Host Species Rabbit
Immunogen REX1 Fusion Protein Ag4334
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314630, 57455, 66932
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.