Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | REX1 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8790
![]() ![]() |
Proteintech Group Inc
135031AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Seems to have no detectable effect on transcription elongation in vitro. Tamura K., Biochem. Biophys. Res. Commun. 309:189-195(2003). Hirosawa M., DNA Res. 6:329-336(1999). The MGC Project Team; Genome Res. 14:2121-2127(2004).Specifications
REX1 | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q7TT28, Q8N1G1 | |
314630, 57455, 66932 | |
REX1 Fusion Protein Ag4334 | |
Primary | |
-20°C | |
REXO1 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
EloA BP1, ELOABP1, Elongin A binding protein 1, KIAA1138, REX1, REXO1, RNA exonuclease 1 homolog, TCEB3BP1 | |
Rexo1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title