Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REXO1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP184863
Description
REXO1 Polyclonal specifically detects REXO1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
REXO1 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
EC 3.1, ELOABP1elongin A-binding protein 1, EloA-BP1REX1, Elongin-A-binding protein 1, KIAA1138elongin A binding protein 1, REX1, RNA exonuclease 1 homolog (S. cerevisiae), TCEB3BP1RNA exonuclease 1 homolog, transcription elongation factor B polypeptide 3 binding protein 1, Transcription elongation factor B polypeptide 3-binding protein 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
REXO1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:PSGKYVVDNSRPPTDLEYDPLSNYSARHLSRASSRDERAAKRPRGSRGSEPYTPAPKKLCDPFGSCDARFSDSEDEAATVPGNEPTT | |
0.1 mL | |
DNA Repair, Editing and Processing Endonucleases | |
57455 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction