Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ RF 9

Click to view available options
:
1mg
Description
RF 9 is a selective NPFF receptor antagonist (Ki values are 58 and 75 nM at hNPFF1 and hNPFF2 receptors respectively). Displays selectivity over several related receptors including NPY Y1 GPR10, GPR54, GPR103 and the opioid receptors. Blocks NPFF-induced hypothermia and increases in atrial blood pressure and heart rate, and prevents opioid-induced tolerance associated with hyperalgesia in vivo.
Specifications
Specifications
| Quantity | 1 mg |
| Product Type | RF 9 |
| Purity | 95% |
| Molecular Weight (g/mol) | 482.62 |
| Inhibitors | RF 9 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction