Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFC4 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB363071 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363071 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363071 Supplier Novus Biologicals Supplier No. NB100403IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RFC4 Polyclonal antibody specifically detects RFC4 in Human samples. It is validated for Western Blot

Specifications

Antigen RFC4
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias A1, A1 37 kDa subunit, Activator 1 37 kDa subunit, Activator 1 subunit 4, MGC27291, replication factor C (activator 1) 4, 37kDa, Replication factor C 37 kDa subunit, replication factor C subunit 4, RFC 37 kDa subunit, RF-C 37 kDa subunit, RFC37replication factor C (activator 1) 4 (37kD)
Host Species Rabbit
Immunogen A 23 amino acid synthetic peptide, which represented a portion of the human Replication Factor C 4, 37 kDa subunit encoded within exon 2 (locusLink ID 5984).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 5984
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.