Learn More
Invitrogen™ RFC4 Polyclonal Antibody

Description
Recommended positive controls: HepG2, LNCAP, MCF7, SKOV3.
Specifications
Specifications
| Antigen | RFC4 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum, PBS with no preservative; pH 7.4 |
| Gene | RFC4 |
| Gene Accession No. | P35249 |
| Gene Alias | A1; A1 37 kDa subunit; Activator 1 37 kDa subunit; activator 1 subunit 4; AI894123; AU040575; EGK_12194; replication factor C (activator 1) 4; replication factor C (activator 1) 4, 37kDa; replication factor C 37 kDa subunit; Replication factor C subunit 4; RFC 37 kDa subunit; RF-C 37 kDa subunit; RFC p37; RFC37; RFC4 |
| Gene Symbols | RFC4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.