Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFC5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15810820UL
Description
RFC5 Polyclonal specifically detects RFC5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RFC5 | |
Polyclonal | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
P40937 | |
RFC5 | |
Synthetic peptides corresponding to RFC5(replication factor C (activator 1) 5, 36.5kDa) The peptide sequence was selected from the N terminal of RFC5. Peptide sequence METSALKQQEQPAATKIRNLPWVEKYRPQTLNDLISHQDILSTIQKFINE. | |
20 μL | |
Cell Cycle and Replication, DNA Repair | |
5985 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Activator 1 36 kDa subunit, Activator 1 subunit 5,36.5 kD subunit, MGC1155, replication factor C (activator 1) 5, 36.5kDa, RF-C 36 kDa subunit, RFC36replication factor C (activator 1) 5 (36.5kD) | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
This product is specific to Subunit or Isoform: 5. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction