Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFP Antibody, Novus Biologicals™
SDP

Catalog No. NBP145840 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145840 0.1 mg
NB018589 0.025 mg
NBP14584005 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP145840 Supplier Novus Biologicals Supplier No. NBP145840

Rabbit Polyclonal Antibody

RFP Polyclonal specifically detects RFP in Non-species specific samples. It is validated for Western Blot.

Specifications

Antigen RFP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 4 ug/mL
Formulation PBS (pH 7.2), 5 mM EDTA, 30% glycerol, 0.5% BSA with 0.03% Proclin 300
Gene Alias dsRedGene Symbol, Recombinant RFP Protein, Red Fluorescent Protein
Gene Symbols DSR
Host Species Rabbit
Immunogen Recombinant RFP expressed from E. coli.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.