Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP1458400.05MG
Description
RFP Polyclonal specifically detects RFP in Non-species specific samples. It is validated for Western Blot.Specifications
RFP | |
Polyclonal | |
Unconjugated | |
PBS containing 30% glycerol, 0.5% BSA with 0.01% Thimerosal | |
Rabbit | |
0.05 mg | |
Non-species specific | |
IgG |
Western Blot | |
0.50 mg/mL | |
Western Blot | |
dsRedGene Symbol, Recombinant RFP Protein, Red Fluorescent Protein | |
Recombinant RFP expressed from E. coli. | |
Primary | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction