Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $402.00
Specifications
Antigen | RFP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018589
![]() |
Novus Biologicals
NBP1458400.025MG |
0.025 mg |
Each for $243.50
|
|
|||||
NBP14584005
![]() |
Novus Biologicals
NBP1458400.05MG |
0.05 mg |
Each for $402.00
|
|
|||||
NBP145840
![]() |
Novus Biologicals
NBP145840 |
0.1 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
RFP Polyclonal specifically detects RFP in Non-species specific samples. It is validated for Western Blot.Specifications
RFP | |
Polyclonal | |
Rabbit | |
dsRedGene Symbol, Recombinant RFP Protein, Red Fluorescent Protein | |
IgG |
Western Blot | |
Unconjugated | |
Non-species specific | |
Recombinant RFP expressed from E. coli. | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title