Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFWD3 Antibody, Novus Biologicals™
SDP

Catalog No. NB10068206 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10068206 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10068206 Supplier Novus Biologicals Supplier No. NB10068206
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RFWD3 Polyclonal specifically detects RFWD3 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen RFWD3
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2 - 5 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_060594.3
Gene Alias EC 6.3.2.-, FLJ10520, ring finger and WD repeat domain 3, RING finger and WD repeat domain-containing protein 3, RING finger protein 201, RNF201E3 ubiquitin-protein ligase RFWD3
Gene Symbols RFWD3
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human ring finger and WD repeat domain 3 using the numbering given in entry NP_060594.3 (GeneID 55159).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 55159
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.