Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFX3 Antibody (OTI3F7), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP273865G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273865G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273865G Supplier Novus Biologicals Supplier No. NBP273865G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RFX3 Monoclonal specifically detects RFX3 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen RFX3
Applications Western Blot
Classification Monoclonal
Clone OTI3F7
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bA32F11.1, DNA binding protein RFX3, MGC87155, Regulatory factor X 3, regulatory factor X, 3 (influences HLA class II expression), transcription factor RFX3
Gene Symbols RFX3
Host Species Mouse
Immunogen Full length recombinant protein of human RFX3 (NP_602304) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5991.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.