Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFX5 Antibody, Novus Biologicals™
SDP

Catalog No. NB100195 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100195 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100195 Supplier Novus Biologicals Supplier No. NB100195
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

RFX5 Polyclonal specifically detects RFX5 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen RFX5
Applications Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, Immunoprecipitation 1 - 4 ug/mg of lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Alias DNA-binding protein RFX5, Regulatory factor X 5, regulatory factor X, 5 (influences HLA class II expression)
Gene Symbols RFX5
Host Species Rabbit
Immunogen Rabbits were immunized with a synthetic peptide, which represented a portion of human Regulatory Factor X 5 encoded within exon 5 (LocusLink ID 5993), conjugated to KLH. After multiple immunizations in Freund's adjuvant, serum was collected. Antibodies were affinity purified using the peptide immobilized on solid support.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 5993
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Gorilla, Chimpanzee and White-tufted-ear marmoset.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.