Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFX7 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP171819FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP171819FR 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP171819FR Supplier Novus Biologicals Supplier No. NBP171819FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RFX7 Polyclonal specifically detects RFX7 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen RFX7
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DNA-binding protein RFX7, FLJ12994, FLJ21104, Regulatory factor X 7, regulatory factor X domain containing 2, Regulatory factor X domain-containing protein 2, regulatory factor X, 7, RFXDC2MGC131836
Gene Symbols RFX7
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1313 and 1363 of human Regulatory Factor X, 7 using the numbering given in entry AAI12937.1 (GeneID 64864).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64864
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.