Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFXANK Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB045916 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045916 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045916 Supplier Novus Biologicals Supplier No. NBP297640AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RFXANK Polyclonal specifically detects RFXANK in Human samples. It is validated for Western Blot.

Specifications

Antigen RFXANK
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ANKRA1Regulatory factor X subunit B, ankyrin repeat-containing regulatory factor X-associated protein, BLS, DNA-binding protein RFXANK, F14150_1, MGC138628, regulatory factor X-associated ankyrin-containing proteinAnkyrin repeat family A protein 1, RFXB, RFX-BRFX-Bdelta4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RFXANK fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 8625
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.