Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGL2 Antibody (OTI10F2), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP273875R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273875R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273875R Supplier Novus Biologicals Supplier No. NBP273875R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RGL2 Monoclonal specifically detects RGL2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen RGL2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI10F2
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias GDS-related protein, HKE1.5, KE1.5, RAB2, member RAS oncogene family-like, RAB2LRas-associated protein RAB2L, ral guanine nucleotide dissociation stimulator-like 2, RalGDS-like 2, RalGDS-like factor
Gene Symbols RGL2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 547-777 of human RGL2 (NP_004752) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5863.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.