Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGP1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB342350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342350 Supplier Novus Biologicals Supplier No. NBP281799MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RGP1 Polyclonal antibody specifically detects RGP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen RGP1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias retrograde Golgi transport protein RGP1 homolog, RGP1 retrograde golgi transport homolog (S. cerevisiae)
Host Species Rabbit
Immunogen RGP1 antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of the human RGP1. The immunogen is located within amino acids 50 - 100 of RGP1. Amino Acid Squence: ALPPPDSSQPDVQPDS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9827
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.