Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGPD5 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241083
Description
RGPD5 Polyclonal specifically detects RGPD5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RGPD5 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
RGPD5 | |
Antibody was raised against a 13 amino acid synthetic peptide from near the amino terminus of human RGPD5. The immunogen is located within amino acids 50 - 100 of RGPD5. Amino Acid Squence: LYELEENTEKAVE | |
0.1 mg | |
Primary | |
Human | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1 μg/mL, ELISA, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL | |
BS63, BS-63, Epididymis Luminal Protein 161, HEL161, Ran-Binding Protein 2-Like 1, Ran-Binding Protein 2-Like 1/2, ranBP2L1, RANBP2L2, RanBP2-Like 1, RanBP2-Like 1/2, RANBP2-Like And GRIP Domain Containing 5, RANBP2-Like And GRIP Domain-Containing Protein 5, RANBP2-Like And GRIP Domain-Containing Protein 5/6, RGP5, RGP6, RGP7, RGPD7, Sperm Membrane Protein BS-63 | |
Rabbit | |
Affinity Purified | |
RUO | |
84220 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction