Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGPD5 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | RGPD5 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
RGPD5 Polyclonal specifically detects RGPD5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RGPD5 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
84220 | |
Antibody was raised against a 13 amino acid synthetic peptide from near the amino terminus of human RGPD5. The immunogen is located within amino acids 50 - 100 of RGPD5. Amino Acid Squence: LYELEENTEKAVE | |
Primary |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Human | |
BS63, BS-63, Epididymis Luminal Protein 161, HEL161, Ran-Binding Protein 2-Like 1, Ran-Binding Protein 2-Like 1/2, ranBP2L1, RANBP2L2, RanBP2-Like 1, RanBP2-Like 1/2, RANBP2-Like And GRIP Domain Containing 5, RANBP2-Like And GRIP Domain-Containing Protein 5, RANBP2-Like And GRIP Domain-Containing Protein 5/6, RGP5, RGP6, RGP7, RGPD7, Sperm Membrane Protein BS-63 | |
RGPD5 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title