Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGPD5 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP241084 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP241084 0.1 mg
NB435997 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP241084 Supplier Novus Biologicals Supplier No. NBP241084
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RGPD5 Polyclonal specifically detects RGPD5 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen RGPD5
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA
Formulation PBS with 0.02% Sodium Azide
Gene Alias BS63, BS-63, Epididymis Luminal Protein 161, HEL161, Ran-Binding Protein 2-Like 1, Ran-Binding Protein 2-Like 1/2, ranBP2L1, RANBP2L2, RanBP2-Like 1, RanBP2-Like 1/2, RANBP2-Like And GRIP Domain Containing 5, RANBP2-Like And GRIP Domain-Containing Protein 5, RANBP2-Like And GRIP Domain-Containing Protein 5/6, RGP5, RGP6, RGP7, RGPD7, Sperm Membrane Protein BS-63
Gene Symbols RGPD5
Host Species Rabbit
Immunogen Antibody was raised against a 23 amino acid synthetic peptide from near the center of human RGPD5. The immunogen is located within amino acids 1130 - 1180 of RGPD5. Amino Acid Squence: DGDAKLERLAAKFKTPELAEEFK
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84220
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.