Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB081025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB081025 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB081025 Supplier Novus Biologicals Supplier No. NBP297095AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RGS1 Polyclonal specifically detects RGS1 in Human samples. It is validated for ELISA.

Specifications

Antigen RGS1
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias 1R20Early response protein 1R20, BL34B-cell activation protein BL34, IER1, IR20immediate-early response 1, B-cell specific, regulator of G-protein signaling 1, regulator of G-protein signalling 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant RGS1 (NP_002913.3; Met1-Lys196).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5996
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.