Learn More
Description
Specifications
Specifications
| Antigen | RGS1 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 405 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | 1R20Early response protein 1R20, BL34B-cell activation protein BL34, IER1, IR20immediate-early response 1, B-cell specific, regulator of G-protein signaling 1, regulator of G-protein signalling 1 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant RGS1 (NP_002913.3; Met1-Lys196). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
