Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220146 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220146 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220146 Supplier Novus Biologicals Supplier No. NBP220146
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RGS1 Polyclonal specifically detects RGS1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RGS1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7), 20% Glycerol with 0.025% Proclin 300
Gene Accession No. Q08116
Gene Alias 1R20Early response protein 1R20, BL34B-cell activation protein BL34, IER1, IR20immediate-early response 1, B-cell specific, regulator of G-protein signaling 1, regulator of G-protein signalling 1
Gene Symbols RGS1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RGS1. The exact sequence is proprietary.
Molecular Weight of Antigen 24 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5996
Test Specificity Human. Species reactivity based on sequence identity: Mouse (86%), Bovine (90%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.