Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS10 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15539620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15539620 20 μL
NBP155396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15539620 Supplier Novus Biologicals Supplier No. NBP15539620UL

Rabbit Polyclonal Antibody

RGS10 Polyclonal specifically detects RGS10 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen RGS10
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q96GN0
Gene Alias regulator of G-protein signaling 10, regulator of G-protein signalling 10
Gene Symbols RGS10
Host Species Rabbit
Immunogen Synthetic peptides corresponding to RGS10 (regulator of G-protein signalling 10) The peptide sequence was selected from the middle region of RGS10. Peptide sequence DQIFNLMKYDSYSRFLKSDLFLKHKRTEEEEEDLPDAQTAAKRASRIYNT.
Molecular Weight of Antigen 21 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6001
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.